missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylcyclohexanol (2-,3-,4- and cis-,trans- mixture) 88.0+%, TCI America™

Supplier:  TCI America M066625ML

Catalog No. M066625ML


Explore available promotions

Description

Description

Specifications

Specifications

Methylcyclohexanol (2-,3-,4- and cis-,trans- mixture)
Yellow
C7H15O
25 mL
114.19
Liquid
25639-42-3
173°C
MFCD01778225
115.20
≥88.0% (GC)

Chemical Identifiers

25639-42-3
C7H15O
115.20
MFCD01778225
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2301

RTECSNumber : GW0175000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions