missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue Chloride, Spectrum™ Chemical

$377.27 - $14956.17

Chemical Identifiers

CAS 7220-79-3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18611770
View Documents
Spectrum Chemical Mfg Cor
ME14525GM
25 g Amber Glass Bottle
Each for $377.27
Only null left
 
18611771
View Documents
Spectrum Chemical Mfg Cor
ME14525KGBL
25 kg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

7220-79-3

Specifications

7220-79-3
Reagent
Blue/Green
Methylene Blue Chloride
1
Amber Glass Bottle
25 g
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.