missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Methylene Blue, pure, certified

Methylene Blue, C16H24ClN3O3S, CAS Number-7220-79-3, Basic Blue 9, 25g, 1.21 to 1.7, Vapor Pressure: Negligible, 27, V,19, 259, CHEBI:6872, Green, 13 (Air - 1.0), 82% min., 200-515-2, 319.85, Pure, Warning, GHS H Statement: Harmful if swallowed.

Supplier:  Thermo Scientific Chemicals 414241000

 View more versions of this product

Catalog No. AC414241000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methylene Blue
7220-79-3
82% min.
MFCD00150008
15, 6132
Solubility in water: 40g/L (20°C). Other solubilities: in methanol: 430g/L
Authentic
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride
6099
663 to 667nm
Pure
Vapor Pressure: Negligible
Green
Fine Crystalline Powder
190.0°C
1.21 to 1.7
C16H24ClN3O3S
27, V,19, 259
Basic Blue 9
XQAXGZLFSSPBMK-UHFFFAOYSA-M
O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C
373.90
CHEBI:6872
319.85
Glass bottle
13 (Air = 1.0)
100 g

Chemical Identifiers

7220-79-3
373.90
XQAXGZLFSSPBMK-UHFFFAOYSA-M
6099
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride
C16H24ClN3O3S
MFCD00150008
Basic Blue 9
CHEBI:6872
O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 200-515-2

RTECSNumber : SO5600000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only