missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylene Blue, Reagent Grade, LabChem™

Supplier:  LabChem LC168508

Catalog No. LC168508


Explore available promotions

Description

Description

Specifications

Specifications

Methylene Blue
93.64,5,1.36
C16H18N3SCl
Passes Test
Reagent
Green
Powder
190°C
7681-11-0,7487-94-7
Soluble in water
319.85
Amber Glass
25 g
SDS
Documents

Documents

Promotions

Promotions