Learn More
![](http://beta-static.fishersci.ca/images/Solvent.gif)
![](http://beta-static.fishersci.ca/images/Blue.gif)
Methylene Chloride, Optima™ for HPLC and GC, Fisher Chemical™
1LT Dichloromethane, for residue analysis, Distol™, stabilised with amylene
$588.00 - $798.67
Chemical Identifiers
CAS | 75-09-2 |
---|---|
Molecular Formula | CH2Cl2 |
Molecular Weight (g/mol) | 84.93 |
MDL Number | MFCD00000881 |
InChI Key | YMWUJEATGCHHMB-UHFFFAOYSA-N |
Synonym | methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm |
PubChem CID | 6344 |
ChEBI | CHEBI:15767 |
IUPAC Name | dichloromethane |
SMILES | ClCCl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
D151-4
|
Thermo Fisher Scientific
BPD1514 |
4 L | Amber Soda Lime Glass |
Each for $588.00
Case of 4 Each for $2,352.00
|
|
||||
D151SK-4
|
Thermo Fisher Scientific
D151SK4 |
4 L | Safe-Cote™ Amber Glass |
Each for $798.67
Case of 4 Each for $3,194.68
|
|
||||
Description
- Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
- Impurity profile is also reported using gas chromatography and three different detectors (ECD, FID, and MS) to demonstrate suitable for different analyses
- Contains approx. 15- 200 ppm amylene as preservative
- Also meets ACS specifications
- Intended for research use only
Chemical Identifiers
75-09-2 | |
84.93 | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
6344 | |
dichloromethane |
CH2Cl2 | |
MFCD00000881 | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
CHEBI:15767 | |
ClCCl |
Specifications
-97°C | |
1 AU max. at 233nm, 0.12 AU max. at 240nm, 0.005 AU max. at 254nm | |
1.33g/cm³ | |
4 L | |
Amber Soda Lime Glass | |
1.42 to 1.423 (at 25°C) | |
99.9% min by GC | |
CH2Cl2 | |
20mmHg at 3502°C | |
0.01% max. | |
YMWUJEATGCHHMB-UHFFFAOYSA-N | |
dichloromethane | |
6344 | |
84.93 | |
High purity mobile phase for HPLC and/or extraction solvent for GC applications |
75-09-2 | |
Colorless | |
39°C | |
Pass Test | |
Liquid | |
1ppm max. | |
0.0002mEq/g max. | |
MFCD00000881 | |
1593 | |
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm | |
ClCCl | |
84.93 | |
CHEBI:15767 | |
≥99.9% | |
Methylene Chloride |
Safety and Handling
![](http://beta-static.fishersci.ca/images/Solvent.gif)
![](http://beta-static.fishersci.ca/images/Blue.gif)
WARNING!
Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. May be harmful if inhaled. May cause irritation of respiratory tract. Inhalation may cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.
NFPA
Health:2
Flammability:1
Instability:0
DOTInformation : DOT Class 6.1, : Poison