missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylenediformamide, 97%, Thermo Scientific Chemicals

$49.00 - $183.03

Chemical Identifiers

CAS 6921-98-8
Molecular Formula C3H6N2O2
Molecular Weight (g/mol) 102.093
MDL Number MFCD00021038
InChI Key QPJQPYQZFKFTHG-UHFFFAOYSA-N
Synonym methylenediformamide, n,n'-methylenebisformamide, n,n'-methylenediformamide, methylenebisformamide, formamide, n,n'-methylenebis, n-formamidomethyl formamide, n,n'-methylenebis formamide, methylene-bis-formamide, n,n-methylenebisformamide, acmc-1b7ve
PubChem CID 81339
IUPAC Name N-(formamidomethyl)formamide
SMILES C(NC=O)NC=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2411603
View Documents
Thermo Scientific Chemicals
B2411603
1 g
Each for $49.00
 
AAB2411606
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
B2411606
5 g
Each for $183.03
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6921-98-8
102.093
QPJQPYQZFKFTHG-UHFFFAOYSA-N
81339
C(NC=O)NC=O
C3H6N2O2
MFCD00021038
methylenediformamide, n,n'-methylenebisformamide, n,n'-methylenediformamide, methylenebisformamide, formamide, n,n'-methylenebis, n-formamidomethyl formamide, n,n'-methylenebis formamide, methylene-bis-formamide, n,n-methylenebisformamide, acmc-1b7ve
N-(formamidomethyl)formamide

Specifications

6921-98-8
C3H6N2O2
1 g
methylenediformamide, n,n'-methylenebisformamide, n,n'-methylenediformamide, methylenebisformamide, formamide, n,n'-methylenebis, n-formamidomethyl formamide, n,n'-methylenebis formamide, methylene-bis-formamide, n,n-methylenebisformamide, acmc-1b7ve
C(NC=O)NC=O
102.093
102.09
Methylenediformamide
141°C to 143°C
MFCD00021038
1811705
QPJQPYQZFKFTHG-UHFFFAOYSA-N
N-(formamidomethyl)formamide
81339
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 230-041-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only