missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylglyoxal, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0215555825

 View more versions of this product

Catalog No. ICN15555825

Explore available promotions

Description

Description

Specifications

Specifications

Methylglyoxal
78-98-8
1.17g/mL
MFCD00006960
methylglyoxal, pyruvaldehyde, pyruvic aldehyde, acetylformaldehyde, acetylformyl, propanal, 2-oxo, 2-ketopropionaldehyde, propanedione, propanolone, methyl glyoxal
CC(=O)C=O
72.06
CHEBI:17158
Liquid
Molecular biology
Colorless
C3H4O2
25 mL
AIJULSRZWUXGPQ-UHFFFAOYSA-N
2-oxopropanal
880
72.06

Chemical Identifiers

78-98-8
72.06
AIJULSRZWUXGPQ-UHFFFAOYSA-N
880
2-oxopropanal
C3H4O2
MFCD00006960
methylglyoxal, pyruvaldehyde, pyruvic aldehyde, acetylformaldehyde, acetylformyl, propanal, 2-oxo, 2-ketopropionaldehyde, propanedione, propanolone, methyl glyoxal
CHEBI:17158
CC(=O)C=O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 201-164-8

Recommended Storage : Store at 4°C, dessicated.

SDS
Documents

Documents

Promotions

Promotions