missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methylmagnesium bromide, 1M solution in THF, AcroSeal™, ACROS Organics™

Supplier:  Thermo Scientific Chemicals 377381000

 View more versions of this product

Catalog No. AC377381000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methylmagnesium bromide
1M Solution in THF
CH3BrMg
MFCD00000041
1.0000g/mL
Solubility in water: vigorous reaction.
C[Mg]Br
119.24
119.24
1
−21°C
Brown-Gray to Brown-Purple
75-16-1,109-99-9
CH3MgBr
0.95 to 1.20M
methylmagnesium bromide, grignard reagent, magnesium, bromomethyl, bromo methyl magnesium, bromo methyl magnesium, unii-22cw9773df, memgbr, memgbr, methymagnesiumbromide, methylmagnesiumbromide, methylmagnesiumbromide
AVFUHBJCUUTGCD-UHFFFAOYSA-M
magnesium;carbanide;bromide
6349
AcroSeal™ Glass bottle
100 mL
Solution

Chemical Identifiers

75-16-1
119.24
AVFUHBJCUUTGCD-UHFFFAOYSA-M
6349
C[Mg]Br
CH3BrMg
MFCD00000041
methylmagnesium bromide, grignard reagent, magnesium, bromomethyl, bromo methyl magnesium, bromo methyl magnesium, unii-22cw9773df, memgbr, memgbr, methymagnesiumbromide, methylmagnesiumbromide, methylmagnesiumbromide
magnesium;carbanide;bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. May cause respiratory irritation. Reacts violently with water. May form explosive peroxides. Suspected of causing cancer. Harmful if swallowed. May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only