Learn More
Methylmagnesium iodide, 3M solution in diethyl ether, AcroSeal™
CAS: 917-64-6 | CH3IMg | 166.24 g/mol
$141.22 - $141.22
Chemical Identifiers
| CAS | 917-64-6 |
|---|---|
| Molecular Formula | CH3IMg |
| Molecular Weight (g/mol) | 166.24 |
| MDL Number | MFCD00001026 |
| InChI Key | AUPXBVDHVRZMIB-UHFFFAOYSA-M |
| Synonym | methylmagnesium iodide, magnesium, iodomethyl, methylmagnesiumiodide, methyl magnesium iodide, iodomethylmagnesium, methylmagnesium iodide solution, 3.0 m in diethyl ether, memgi, ch3mgi, methyl-magnesium iodide |
| PubChem CID | 101907 |
| IUPAC Name | magnesium;carbanide;iodide |
| SMILES | C[Mg]I |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC427401000
|
Thermo Scientific Chemicals
427401000 |
100 mL | AcroSeal™ Glass bottle |
Each for $141.22
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 917-64-6 | |
| 166.24 | |
| AUPXBVDHVRZMIB-UHFFFAOYSA-M | |
| 101907 | |
| C[Mg]I |
| CH3IMg | |
| MFCD00001026 | |
| methylmagnesium iodide, magnesium, iodomethyl, methylmagnesiumiodide, methyl magnesium iodide, iodomethylmagnesium, methylmagnesium iodide solution, 3.0 m in diethyl ether, memgi, ch3mgi, methyl-magnesium iodide | |
| magnesium;carbanide;iodide |
Specifications
| 917-64-6,60-29-7 | |
| −40°C | |
| CH3IMg | |
| MFCD00001026 | |
| 1.26 | |
| AUPXBVDHVRZMIB-UHFFFAOYSA-M | |
| magnesium;carbanide;iodide | |
| 101907 | |
| Methylmagnesium iodide |
| 1.2600g/mL | |
| AcroSeal™ Glass bottle | |
| CH3MgI | |
| 100 mL | |
| methylmagnesium iodide, magnesium, iodomethyl, methylmagnesiumiodide, methyl magnesium iodide, iodomethylmagnesium, methylmagnesium iodide solution, 3.0 m in diethyl ether, memgi, ch3mgi, methyl-magnesium iodide | |
| C[Mg]I | |
| 166.24 | |
| 166.24 |
Safety and Handling
GHS H Statement
Harmful if swallowed. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. In contact with water releases flammable gases which may ignite spontaneously. Extremely flammable liquid and vapor. May form explosive peroxides. Reacts violently with water. Repeated exposure may cause skin dryness or cracking.
GHS P Statement
Wear eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid breathing dust/fume/gas/mist/vapors/spray. Store in a dry place. Store in a closed container. Keep container tightly closed.
GHS Signal Word: Danger
EINECSNumber : 213-031-1
RUO – Research Use Only