missing translation for 'onlineSavingsMsg'
Learn More

Methylmaleic Hydrazide 98.0+%, TCI America™

$145.95 - $446.25

Chemical Identifiers

CAS 5754-18-7
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.12
MDL Number MFCD00067746,MFCD00023867
InChI Key QAVYOWFNXMHVEL-UHFFFAOYSA-N
Synonym 4-methylpyridazine-3,6-diol, 3,6-dihydroxy-4-methylpyridazine, 1,2-dihydro-4-methyl-3,6-pyridazinedione, 4-methyl-3,6-pyridazinediol, 3,6-pyridazinedione, 1,2-dihydro-4-methyl, 3, 1,2-dihydro-4-methyl, 2-butenedioic acid, cyclic hydrazide, z, acmc-1aqja, 4-methylmaleic hydrazide, methylmaleic acid hydrazide
PubChem CID 79826
IUPAC Name 4-methyl-1,2,3,6-tetrahydropyridazine-3,6-dione
SMILES CC1=CC(=O)NNC1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M21825G
View Documents
TCI America
M21825G
5 g
Each for $145.95
Only null left
 
M218225G
View Documents
TCI America
M218225G
25 g
Each for $446.25
Only null left
 
Description

Description

Specifications

Chemical Identifiers

5754-18-7
126.12
QAVYOWFNXMHVEL-UHFFFAOYSA-N
79826
CC1=CC(=O)NNC1=O
C5H6N2O2
MFCD00067746,MFCD00023867
4-methylpyridazine-3,6-diol, 3,6-dihydroxy-4-methylpyridazine, 1,2-dihydro-4-methyl-3,6-pyridazinedione, 4-methyl-3,6-pyridazinediol, 3,6-pyridazinedione, 1,2-dihydro-4-methyl, 3, 1,2-dihydro-4-methyl, 2-butenedioic acid, cyclic hydrazide, z, acmc-1aqja, 4-methylmaleic hydrazide, methylmaleic acid hydrazide
4-methyl-1,2,3,6-tetrahydropyridazine-3,6-dione

Specifications

5754-18-7
White-Yellow
MFCD00067746,MFCD00023867
4-methylpyridazine-3,6-diol, 3,6-dihydroxy-4-methylpyridazine, 1,2-dihydro-4-methyl-3,6-pyridazinedione, 4-methyl-3,6-pyridazinediol, 3,6-pyridazinedione, 1,2-dihydro-4-methyl, 3, 1,2-dihydro-4-methyl, 2-butenedioic acid, cyclic hydrazide, z, acmc-1aqja, 4-methylmaleic hydrazide, methylmaleic acid hydrazide
CC1=CC(=O)NNC1=O
126.12
126.12
Crystalline Powder
287°C
C5H6N2O2
5 g
QAVYOWFNXMHVEL-UHFFFAOYSA-N
4-methyl-1,2,3,6-tetrahydropyridazine-3,6-dione
79826
≥98.0% (GC)
Methylmaleic Hydrazide
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'