missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Metro™ MetroMax i Basket Glassware Cart

Store or move your glassware around the lab with confidence. MetroMax I Basket Glassware Cart is a Level 2 cart with built-in Microban antimicrobial product protection that allows you to configure its arrangement, ensuring storage and movement won't disrupt your workday.

Supplier:  Metro™ GWBX31

 View more versions of this product

Catalog No. 13-451-091


Explore available promotions

Description

Description

Modular design offers ultimate flexibility. Reinforced polymers provide superior strength (up to 800 lb. per shelf).

  • Corrosion-proof, suitable for damp and humid conditions
  • Lifetime warranty against rust and corrosion
  • Shelves wipe down easily and have a retaining edge that contains drips and spills
  • Two-basket and four-basket capacity glassware solutions are offered in a spacesaving 18 x 36 in. (457 x 914mm) footprint
  • Solid MetroMax bottom shelf provides spill retention and can be removed from shelf frame for easy cleaning
  • Wide range of accessoriesavailable separately) including all-polymer casters for maximum corrosion resistance and premium performance

Order Info

Casters not included (5PC Series Metro casters are recommended for superior lifetime performance).

Specifications

Specifications

Glassware Cart
36 x 18 x 4 in. (91.4 x 45.7 x 10.2 cm)
40 in.
Solid MetroMax i bottom shelf
91.4 cm
362.8 kg
18 in.
2-Basket Unit
Glassware transportation
101.6 cm
36 in.
800 lb.
Polymer
45.7 cm
SDS
Documents

Documents

Promotions

Promotions