missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Metro™ MW Series Utility Cart with Solid and Wire Shelves
Made of Super Erecta™ shelves with plastic split sleeves, two one-piece handles and casters with donut bumpers, Metro MW Series Utility Cart with Solid and Wire Shelves make a smart addition to any workplace.
Supplier: Metro™ MW402
Description
Stainless steel, chrome shelf and handle material.
Cart
- Load capacity: 375 lb. (170.1kg)
- Fully interchangeable Super Erecta™ shelves
- Center shelf can be positioned at any height in 1 in. (2.5cm) increments
- One-piece stainless-steel or chrome handles (depends on model) add rigidity and durability
- 18 in. (46cm) carts have 4 in. dia. (10cm) casters; 21 in. (53cm) and 24 in. (61cm) wide carts have 5 in. dia. (13cm) casters
- Three tiers
- 375 lbs (170kg) weight capacity per cart
- 18-gauge stainless-steel, doubly thick at the edges for rigidity and strength
- Raised edges help contain spills and make cleanup easier
- Sturdy corner castings facilitate mounting
- Chrome-plated or stainless-steel shelves available with trademarked truss design for added strength
- Corners slide easily over special tapered snap-together plastic sleeves to fasten them securely in place
Order Info
Shipped knocked down. Assembles/disassembles quickly and easily without tools.
Specifications
Utility Cart | |
3-tier Model | |
Transporting materials that require an expanse of flat, solid surface | |
39 in. | |
Super Erecta™ 2 Chrome plated wire shelves and one Stainless Steel solid (flat) shelf with Plastic split sleeves, 2 one-piece Chrome plated handles, 4 in. dia. caster, 3.5 in. donut bumpers | |
45.72 cm | |
170 kg | |
Stainless Steel, Chrome | |
47 lb. | |
76.2 cm |
NSF | |
18 x 30 x 39 in. | |
Chrome | |
99 cm | |
18 in. | |
375 lb. | |
3 | |
21.3 kg | |
30 in. |