missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Metro™ Starsys™ Overhead Storage Unit

Microban™ antimicrobial protection prevents growth of stains and odor-causing bacteria.

Supplier:  Metro™ SXRDOH27P2S

 View more versions of this product

Catalog No. 13-452-505


Explore available promotions

Description

Description

  • Built on same widths as Starsys base units utilizing very similar construction
  • Insures that wall cabinets align with lower product configuration in application
  • All overhead cabinets include corrugated polymer inner panels in their assembly as well as clean-design back panel, molded with cosmetic side facing inside of cabinet, creating much cleaner appearance - especially when used without doors or in conjunction with clear door
  • Necessary wall mount rails/hardware are included with each wall cabinet
  • Microban™ antimicrobial protection inhibits growth of stain and odor-causing bacteria on Starsys, keeping surface areas cleaner between cleanings
Specifications

Specifications

Overhead Storage Unit
41.3 in.
Polymer
41.3 in.
13.8 in.
Double
2
29.5 in.
29.5 in.
Solid/no view doors, double width
13.8 x 41.3 x 29.5 in.
Taupe
None
Videos
SDS
Documents

Documents

Promotions

Promotions