Metro™ Super Erecta™ Wire Shelf, Stainless Steel Finish
Wire shelves in stainless steel finish allow adjustable configurations when used with Metro Super Erecta shelving systems.
Supplier:  Metro™ 1472NS
 View more versions of this product
Catalog No. 19-030-973
Explore available promotions
Description
Description
  • Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
  • Type 304 polished stainless steel
  • Used for the most aggressive applications and environments
  • Stands up to high temperature automated cart wash and autoclave applications
Specifications
Specifications
35.5 x 182.9 in. (14 x 72 cm)
Wire Shelf
355 mm
Stainless Steel
Plastic sleeves and wedges
14 in.
Metro™ Super Erecta™ shelving system