Metro™ Super Erecta™ Wire Shelf, Stainless Steel Finish
Wire shelves in stainless steel finish allow adjustable configurations when used with Metro Super Erecta shelving systems.
Supplier:  Metro™ 2160NS
 View more versions of this product
Catalog No. 19-030-979
Explore available promotions
Description
Description
  • Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
  • Type 304 polished stainless steel
  • Used for the most aggressive applications and environments
  • Stands up to high temperature automated cart wash and autoclave applications
Specifications
Specifications
53 x 152.4 in. (21 x 60 cm)
Wire Shelf
530 mm
Stainless Steel
Plastic sleeves and wedges
21 in.
Metro™ Super Erecta™ shelving system