Metro™ Super Erecta™ Wire Shelf, Stainless Steel Finish
Wire shelves in stainless steel finish allow adjustable configurations when used with Metro Super Erecta shelving systems.
Supplier: Metro™ 3072NS
Description
- Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
- Type 304 polished stainless steel
- Used for the most aggressive applications and environments
- Stands up to high temperature automated cart wash and autoclave applications
Specifications
76 x 182.9 in. (30 x 72 cm) | |
Wire Shelf | |
760 mm | |
Stainless Steel |
Plastic sleeves and wedges | |
30 in. | |
Metro™ Super Erecta™ shelving system |