missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Metronidazole, 99%

Metronidazole, CAS# 443-48-1, is a member of the class of imidazoles with limited antibiotic activity that acts against protozoa, anaerobic gram-positive, and anaerobic gram-negative bacteria.

Supplier:  Thermo Scientific Chemicals 210340050

 View more versions of this product

Catalog No. AC210340050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• Metronidazole is a synthetic nitroimidazole derivative with antiprotozoal and antibacterial activities and acts against protozoa, anaerobic gram-positive, and anaerobic gram-negative bacteria
• Metronidazole is considered a prodrug that produces nitroso-containing intermediates in the cell that can covalently bind DNA, disrupting helical structure, inducing DNA strand breaks, and inhibiting bacterial nucleic acid synthesis, that results in bacterial cell death

Applications

• Metronidazole shows anti-parasitic and anti-amoebic activities.
• It has been used to prevent the growth of gram-negative anaerobes belonging to the Bacteroides and Fusobacterium, and gram-positive anaerobes such as Peptostreptococcus and Clostridia. This compound affects a high percentage of gram-negative bacteria and has higher tissue penetration that gives it an advantage compared to other compounds.
• This compound was used initially to suppress protozoans such as Entamoeba histolytica, Giardia lamblia, and Trichomonas vaginalis.
• It has been commonly applied for the eradication of Helicobacter pylori.
• It also has a radio-sensitizing effect on hypoxic tumor cells and can have a mutagenic activity in in vivo animal models.

Specifications

Specifications

Metronidazole
98.5
159°C to 163°C
Authentic
Glass bottle
MFCD00009750
15, 6236
Solubility in water: 1g/100mL (20°C). Other solubilities: soluble in diluted acids,0.5g/100mL ethanol- <0.05g/100mL ether (20°C),<0.05g/100mL chloroform (20°C),sparingly soluble in dmf
CC1=NC=C(N1CCO)[N+](=O)[O-]
171.16
CHEBI:6909
99%
443-48-1
100.0
White to Yellow
99%
C6H9N3O3
5 g
metronidazole, metronidazol, flagyl, 2-methyl-5-nitroimidazole-1-ethanol, anagiardil, trichazol, gineflavir, meronidal, metronidaz, novonidazol
VAOCPAMSLUNLGC-UHFFFAOYSA-N
2-(2-methyl-5-nitroimidazol-1-yl)ethanol
4173
171.16
Crystalline Powder

Chemical Identifiers

443-48-1
171.16
VAOCPAMSLUNLGC-UHFFFAOYSA-N
4173
2-(2-methyl-5-nitroimidazol-1-yl)ethanol
C6H9N3O3
MFCD00009750
metronidazole, metronidazol, flagyl, 2-methyl-5-nitroimidazole-1-ethanol, anagiardil, trichazol, gineflavir, meronidal, metronidaz, novonidazol
CHEBI:6909
CC1=NC=C(N1CCO)[N+](=O)[O-]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause cancer.

GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.

GHS Signal Word: Danger

EINECSNumber : 207-136-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only