Mettler Toledo™ Adapters for pH Meters
Accessory adapters
Supplier:  Mettler Toledo™ 51302329
 View more versions of this product
Catalog No. 01-910-541
Explore available promotions
Description
Description
LTW-MiniDin Conductivity Adapter
  • The appropriate cable to connect the conductivity sensor mini DIN to a portable meter.  Adapter is required for the following meters: FiveGo, Seven2Go, Seven2Go Pro, SevenGo Duo, SevenGo Duo Pro.
Specifications
Specifications
LTW-MiniDin Conductivity Adapter
FiveGo, Seven2Go, Seven2Go Pro, SevenGo Duo, SevenGo Duo Pro