missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mettler Toledo™ Analytical Individual Weights: ASTM Class 2, with Certificate

Low magnetic permeability and high corrosion resistance for reliable stable values

$225.55 - $2452.92

Specifications

Certifications/Compliance With certificate, ISO17025:2005, ISO9001, ISO14001 environmental standard
Classification ASTM Class 2
Material Stainless Steel
Density 7.9kg/dm3 at 20°C
Includes Plastic box
View More Specs

Products 17
Catalog Number Mfr. No. Mass Product Type Shape Price Quantity  
Catalog Number Mfr. No. Mass Product Type Shape Price Quantity  
01-915-786
View Documents
Mettler Toledo™
11123526
5 g Two-Piece Analytical Weight With Knob Cylindrical with Handling Knob
Each for $262.03
 
01-915-787
View Documents
Mettler Toledo™
11123527
10 g Two-Piece Analytical Weight With Knob Cylindrical with Handling Knob
Each for $279.25
 
01-915-788
View Documents
Mettler Toledo™
11123528
20 g Two-Piece Analytical Weight With Knob Cylindrical with Handling Knob
Each for $287.85
 
01-915-789
View Documents
Mettler Toledo™
11123529
50 g Two-Piece Analytical Weight With Knob Cylindrical with Handling Knob
Each for $290.72
 
01-915-791
View Documents
Mettler Toledo™
11123530
100 g Two-Piece Analytical Weight With Knob Cylindrical with Handling Knob
Each for $308.89
 
01-915-793
View Documents
Mettler Toledo™
11123531
200 g Two-Piece Analytical Weight With Knob Cylindrical with Handling Knob
Each for $516.40
 
01-915-794
View Documents
Mettler Toledo™
11123532
500 g Two-Piece Analytical Weight With Knob Cylindrical with Handling Knob
Each for $621.60
 
01-915-795
View Documents
Mettler Toledo™
11123533
1 kg Two-Piece Analytical Weight With Knob Cylindrical with Handling Knob
Each for $1,157.14
 
01-915-796
View Documents
Mettler Toledo™
11123534
2 kg Two-Piece Analytical Weight With Knob Cylindrical with Handling Knob
Each for $1,668.75
 
01-915-797
View Documents
Mettler Toledo™
11123535
5 kg Two-Piece Analytical Weight With Knob Cylindrical with Handling Knob
Each for $2,452.92
 
01-915-780
View Documents
Mettler Toledo™
11123520
50 mg Analytical Sheet Weight Square
Each for $226.65
 
01-915-781
View Documents
Mettler Toledo™
11123521
100 mg Analytical Sheet Weight Square
Each for $225.55
 
01-915-783
View Documents
Mettler Toledo™
11123523
500 mg Analytical Sheet Weight Square
Each for $226.65
 
01-915-784
View Documents
Mettler Toledo™
11123524
1 g Two-Piece Analytical Weight With Knob Cylindrical with Handling Knob
Each for $257.25
 
01-915-775
View Documents
Mettler Toledo™
11123515
1 mg Analytical Sheet Weight Square
Each for $235.48
 
01-915-776
View Documents
Mettler Toledo™
11123516
2 mg Analytical Sheet Weight Square
Each for $235.48
 
01-915-778
View Documents
Mettler Toledo™
11123518
10 mg Analytical Sheet Weight Square
Each for $226.65
 
Description

Description

Features
 
  • Premium stainless steel
  • State-of-the-art turning procedures, improved coatings, stronger substrates
  • Electrolytic polishing ensures smooth, glossy surface
  • Magnetization and susceptibility of all weights are strictly controlled to ensure compliance with standards
 
Construction
  • Two-piece screw-knob construction
  • 10mg to 20mg weights are sheet weights made of stainless steel
  • 200 to 500mg weights are sheet weights made of steel
  • 1g to 1kg weights are cylindrical knob weights made of steel
  • Density: 7.9kg/dm3

Includes: Weight, plastic box, and certificate.
Specifications

Specifications

With certificate, ISO17025:2005, ISO9001, ISO14001 environmental standard
Stainless Steel
Plastic box
1
ASTM Class 2
7.9kg/dm3 at 20°C
Each
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'