missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mettler Toledo™ Redox Buffer Solutions

Storage and refill solutions for redox electrodes.

$117.50 - $123.95

Specifications

Type Buffer Solution
Includes Quality inspection certificate
Water Soluble Completely Miscible
View More Specs

Products 3
Catalog Number Mfr. No. pH Range Filling Solution Type Packaging mV Range Price Quantity  
Catalog Number Mfr. No. pH Range Filling Solution Type Packaging mV Range Price Quantity  
01-913-237
View Documents
Mettler Toledo™
51350060
7 Redox Buffer; UH = 427mV 250 mL Bottle 220 mV
Each for $117.50
 
01-915-638 DFS Item
View Documents
Mettler Toledo™
51350062
7 Redox Buffer; UH = 427mV 6 x 250 mL 220 mV N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
01-915-637
View Documents
Mettler Toledo™
51350064
0.1 Redox Buffer; UH/pH: 0.1 6 x 30 mL Bottle 468 mV
Pack of 6 Each for $123.95
 
Description

Description

Includes a quality inspection certificate which guarantees the stated values and traceability.

Certifications

NIST traceable

Specifications

Specifications

Buffer Solution
Quality inspection certificate
Completely Miscible
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'