Mettler Toledo™ CarePacs™: Customized Third Weight
Third weight for customer-specific testing (e.g., minimum weight determination)
Supplier: Mettler Toledo™ 11123150
Description
- Optional customized third weight for Mettler Toledo CarePacs
- ASTM or OIML certified
Specifications
Customized 3rd Weight | |
ASTM Class 1 | |
100 mg | |
Without certificate | |
1 |
ISO9001, ISO14001 environmental standard | |
Stainless Steel | |
100 mg | |
Each |