Learn More
Mettler Toledo™ CarePacs™: Customized Third Weight
Third weight for customer-specific testing (e.g., minimum weight determination)
Supplier:  Mettler Toledo™ 11123056
 View more versions of this product
Catalog No. 01-912-144
Only null left
Explore available promotions
Description
Description
  • Optional customized third weight for Mettler Toledo CarePacs
  • ASTM or OIML certified
Specifications
Specifications
Customized 3rd Weight
OIML Class E2
10 g
Without certificate
1
ISO9001, ISO14001 environmental standard
Stainless Steel
10 g
Each
Product Suggestions
Product Suggestions