Conductivity Standards for METTLER TOLEDO Conductivity Probes, METTLER TOLEDO™
Recommended NIST traceable solutions for calibrating or verifying METTLER TOLEDO conductivity electrodes.
Supplier: METTLER TOLEDO 51302049
Description
- For daily verification: to monitor health of electrode and ensure measurement accuracy
- For initial calibration of nominal cell constant electrodes
- Be sure to screw cap tightly immediately after use (sachets minimize the chance of error due to environmental interaction)
- Conductivity standards subject to change values when exposed to environment; water evaporation raises conductivity values
Specifications
30 x 20 mL | |
Conductivity Standard | |
Conductivity Standard | |
1413 μS/cm |
20 mL | |
Yes | |
Conductivity Electrodes |