Mettler Toledo™ DL31 Karl Fischer™ Titrator Replacement Parts: Electrodes
Replacement parts for DL31 Karl Fischer titrators.
Supplier: Mettler Toledo™ 89598
Description
Combined platinum ring redox electrode with a ceramic frit for redox titrations with change of the pH value.Specifications
Combined Platinum Ring Electrode | |
Ceramic frit | |
149.5 mm | |
0°C to 80°C |
For redox titrations | |
5.8 in. | |
0°F to 176°F |