Mettler Toledo™ DO308 Karl Fischer™ Drying Oven
Allows easy and accurate water determination of solid or viscous samples. Mettler Toledo Drying Ovens are for use with volumetric or coulometric Karl Fischer titrators.
Supplier:  Mettler Toledo™ 51371200
Catalog No. 01911241
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Includes: All necessary parts including the drying unit with gas flow measurement. If purging with dry air is desired, an optional air pump is available. Aluminum inserts sold separately (01-912-515).
Description
Description
Easy and fast manual handling
  • Easy handling for left- and right-handed operators
  • Sample exchange and sample transport into the heated zone of the oven is provided for by simple transport system with magnetic guide rod attached to glass sample boat
  • Equilibrium conditions are quickly reached
  • Cold and hot zone can be purged separately with dry carrier gas using stopcock
  • Ambient moisture introduced during sample exchange or addition via sleeve opening is quickly carried away; stable drift is reached within short period of time
Enhanced operational safety
  • No need to remove transfer tube from titration cell after switching off the oven; just open stopcock to avoid contamination of oven due to underpressure
  • Different temperature ranges can be preprogrammed in one cycle
  • Safety is further improved by automatic shutdown after analysis is completed
Specifications
Specifications
Karl Fischer Drying Oven
Accurate water determination of solid or viscous samples
500 mm
40°C to 300°C; ±5°C at 300°C
9.1 kg
Drying Oven
All necessary parts including the drying unit with gas flow measurement. If purging with dry air is desired, an optional air pump is available
Footprint housing: 256 x 180mm
100/230 V
220 mm