Mettler Toledo™ Draft Chamber Kits
Plexiglas™ shields
$911.51 - $1066.15
Specifications
For Use With (Equipment) | AX, UMX2, MX5, AT, UMT2, MT5, AG, AB-S, PR, SR, PG |
---|---|
Type | Draft Chamber Kit |
Catalog Number | Mfr. No. | Dimensions (L x W x H) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W x H) | Price | Quantity | |||||
01-910-68
|
Mettler Toledo™
63052324 |
18 x 25 x 25 in. (46 × 64 × 64cm) |
Each for $1,066.15
|
|
|||||
09-313-152F
|
Mettler Toledo™
63052337 |
16 x 16 x 17 in. (41 × 41 × 43cm) |
Each for $911.51
|
|
|||||
Description
Can be assembled to fit around any Mettler Toledo Series balance (09-313-152F will not fit SR Series balances).
Specifications
AX, UMX2, MX5, AT, UMT2, MT5, AG, AB-S, PR, SR, PG | |
Draft Chamber Kit |