missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
METTLER TOLEDO™ EasyPlus Easy pH Titrator
Used to automate and speed up simple acid/base titrations. The EasyPlus Easy pH Titrator is ideal for the determining the acidity content of beverages, alkalinity of cleaning reagents and chemicals, free fatty acid content of cooking oils, and alkalinity (m-value) and acidity (p-value) of water.
Supplier: METTLER TOLEDO™ 30060041
Includes: Standard delivery components:
- EG11-BNC sensor pH aqueous
- EasyPlus burette 20mL
- EasyPlus tubing set instrument
- EasyStir GT
- EasyPlus titration head GT
- EasyPlus drying tube
- Valve
- Bottle head
- Insert set titration head GT
- Burette cover set
Description
- Swiss design and construction in accordance with highest software and hardware standards
- High quality component construction, capable of enduring harsh laboratory conditions
- Small design capable of performing any basic titration with ease
- Built-in iTitrate™ intelligence requires few parameters to be set before the instrument is ready for titration
- Gives accurate, reproducible and repeatable results
- Shows results automatically which are stored and are available for print or transfer to EasyDirect™ software
- Smartphone-style Apps design is used to control all major instrument functions making it easy to use
- LongClick™ helps to start the last analysis run without always navigating the routine analyses
- EasySetup guides steps for installation and programming
- Simple and intuitive navigation speaks 14 different languages including Arabic, Chinese, Thai, Portuguese and English
- Reduces training time with the help of App style user interface while increasing work speed and simplicity
- Meets all acid/base titration needs
Specifications
EasyPlus burette 20 mL (included) | |
0.01 | |
Yes | |
10 mL, 20 mL (Standard Delivery), or 50 mL | |
4.10/9.21/11.00; 1.68/4.00/10.01; 1.680/4.003/12.460; or User defined | |
BNC, USB A, USB B, Mini-DIN, RCA | |
Export via USB, USB Printer | |
4.3 inch, 480 x 272 pixel RGB | |
Detection of up to 3 | |
EG11-BNC sensor pH aqueous, EasyPlus burette 20mL, EasyPlus tubing set instrument, EasyStir GT, EasyPlus titration head GT, EasyPlus drying tube, valve, bottle head, insert set titration head GT, burette cover set | |
Touchscreen | |
PP GF30 | |
Easy pH | |
0.5% RSD | |
0 to 14 | |
±0.1mV | |
0.1°C | |
User Defined | |
41 to 104°F | |
5 to 40°C | |
Yes | |
iTitrate | |
Yes | |
12 months | |
6.7 in. | |
8.66 in. | |
100 to 240V ±10% 50/60Hz | |
50/60 Hz |
0.1 | |
0.2°C | |
Variable or fixed value | |
2 | |
CE, CSA | |
Maximum number of samples in a series is 5. When choosing this option the mean value and RSD is calculated. | |
Easy pH | |
EasyPlus Automated Titrator | |
Potentiometric - acid/base titrations | |
BNC, USB A, USB B, Mini-DIN, RCA | |
15 | |
Acid/base endpoint or inflection/equivalence point titrations | |
24 VDC / 1.25 A | |
±2000 mV | |
Max. 80% at 31°C | |
±0.1mV | |
None, Fixed, Variable | |
Low, Medium, High, Maximum | |
0 to 100°C | |
°C / °F | |
Normal, Fast, Cautious, Very cautious, User defined | |
Direct, back compensated, blank compensated | |
100 mL Beaker | |
220 x 170 x 350 mm | |
170 mm | |
220 mm | |
100 to 240V ±10% |