missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mettler Toledo™ Accessories for EasyPlus™ Titrator

For use with Mettler Toledo EasyPlus Titrators.

Supplier:  Mettler Toledo™ 30043900

 View more versions of this product

Catalog No. 01-912-531

Explore available promotions

Description

Description

  • Volume: 10mL
Specifications

Specifications

EasyPlus Burette (10 mL)
10 mL
Videos
SDS
Documents

Documents

Promotions

Promotions