Mettler Toledo™ G20 Potentiometric Titrator for Food, Water, and Chemical Titrations
Simple, dependable, and ready-to-go. Mettler Toledo G20 Potentiometric Titrator comes with One Click™ user interface with user-specific homescreen, shortcuts, and operating language.
Supplier:  Mettler Toledo™ 51105520
Catalog No. 09-317-580
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Plug and Play buret and sensor recognition
  • Compact and simple automation with the Rondolino titration stand (01-912-343 only)
  • Use the Solvent Manager to avoid contact with harmful chemicals
  • GLP compliant Sensor test promises reliable results
  • LabX™ light titration offers data security and comprehensive documentation
Note: Electrode is required for use (sold separately).
Specifications
Specifications
TFT
Food, Water and chemical titrations
±2000 mV
9.05 in.
8.2 in.
13.1 in.
Potentiometric Titrator
G20 Titrator with a 20mL burette. (Electrode sold separately.)
9.05 x 8.2 in. (320 x 210mm)
320 mm
210 mm
333 mm