Mettler Toledo™ Glass Cylinder For Titration Sensor Interchangeable Burette
Replacement part for DV1005 or DV1020 titration burettes.
Supplier:  Mettler Toledo™ 51107100
 View more versions of this product
Catalog No. 01-911-103
Explore available promotions
Description
Description
Replacement part for DV1005 or DV1020 titration burettes.
Compliance
ISO 8655-3
Specifications
Specifications
Cylinder
Glass
5mL titration sensor interchangable burettes