Mettler Toledo™ Halogen Moisture Analyzer Glass Fiber Discs
Mettler Toledo filters are the only filters that are officially supported by Mettler Toledo Moisture Analyzers for optimal use and guaranteed performance.
Supplier: Mettler Toledo™ 00214464
Description
For use with samples with high moisture content. The absorbent glass fiber filter is beneficial for liquid or pasty substances. For a further reduction in analysis time.
Specifications
Glass | |
Moisture Disc | |
88.9 mm |
Round | |
Mettler Toledo Halogen Moisture Analyzer |