missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
METTLER TOLEDO™ InLab™ Viscous pH Electrodes
A glass pH electrode with an integrated temperature sensor featuring the SteadyForce™ reference system, which is pressurized to prevent the junction from clogging.
$1498.75 - $1897.60
Specifications
Junction | Ceramic |
---|---|
Material | Glass |
pH Range | 0 to 14 |
Temperature (Metric) Operating | 0°C to 130°C |
For Use With (Application) | Highly viscous, highly glutinous samples |
Catalog Number | Mfr. No. | Model | Type | Connector Types | For Use With (Equipment) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Type | Connector Types | For Use With (Equipment) | Price | Quantity | |||
01911470
|
METTLER TOLEDO™
51343150 |
InLab™ Viscous | Combination Electrode | S7 | S7 to BNC (30281915) |
Each for $1,498.75
|
|
|||
01-911-440
|
METTLER TOLEDO™
51343151 |
InLab™ Viscous Pro-ISM | Combination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology | Multi-Pin | Multi-Pin to BNC/RCA (30281896) |
Each for $1,897.60
|
|
|||
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Cosmetics
- Gels, soaps, shampoos
- Resins
- Paints
- Varnishes and glues
- Colorants and dyes
- Oily samples
- TRIS compatible
Accessories & Care
- Ceramic Junction and non-refillable - no additional electrolyte required
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications
Ceramic | |
0 to 14 | |
Highly viscous, highly glutinous samples | |
HA Glass | |
32°F to 266°F | |
1.57 in. |
Glass | |
0°C to 130°C | |
SteadyForce | |
3 mol/L KCl | |
0°C to 100°C | |
40 mm |