missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mettler Toledo™ ISA Solutions

Solutions for perfectION™ combined ion selective electrodes

Supplier:  Mettler Toledo™ 51344761

 View more versions of this product

Catalog No. 01-928-351


Explore available promotions

Description

Description

Solutions (ionic-strength adjustment buffers)

  • Ion Electrolyte B (for chloride, copper, iodide, silver/sulfide ISE's), 475mL
  • ISA for calcium electrode, 475mL
  • ISA for potassium electrode, 475mL
  • ISA for nitrate electrode, 475mL
  • Nitrate ISS (for suppressing interference), 475mL
  • Fluoride TISAB II with CDTA, 3790mL
  • Fluoride TISAB III with CDTA (concentrate), 475mL

Specifications

Specifications

perfectION calcium ISA
Calcium
475 mL
ISA Solution
475 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions