Mettler Toledo™ LabX™ Balance Express Software
Weighs different classes of material with improved documentation allowing electronic integration of weights and sample IDs with other software systems within lab.
Supplier:  Mettler Toledo™ 11153220
 View more versions of this product
Catalog No. 01-909-387
Explore available promotions
Description
Description
  • Express edition
  • Connect one instrument with option to up to 3 balances
  • Graphical method editor supports any weighing application
  • Built in method library for quick weighing method development
  • Comprehensive and customizable report editor
LabX seamlessly integrates with third-party systems such as LIMS, ELN, SAP and CDS
Specifications
Specifications
Excellence XP or XS Balance
Connect additional balance to the LabX™ Expr
Express Edition License: for 1 Excellence XP or XS
Balance Software