Mettler Toledo™ Moisture Analyzer Sample Pans
Mettler Toledo sample pans are the only sample pans that are officially supported by Mettler Toledo Moisture Analyzers for optimal use and guaranteed performance.
$216.49 - $425.82
Specifications
Shape Round
Type Moisture Pan
For Use With (Equipment) All Mettler Toledo Moisture Analyzers
Diameter (Metric) 88.9 mm
View More Specs
Products 3
Catalog Number Mfr. No. Material For Use With (Equipment) Description Price Quantity  
Catalog Number Mfr. No. Material For Use With (Equipment) Description Price Quantity  
01-909-427
View Documents
Mettler Toledo™
00214462
Stainless Steel All Mettler Toledo Moisture Analyzers Reusable Pans - Pk of 3
Each for $425.82
01-913-97B
View Documents
Mettler Toledo™
13954
Steel All Mettler Toledo Moisture Analyzers Reusable Pans - High edges (15mm)
Each for $216.49
01-913-97A Edge
View Documents
Mettler Toledo™
13865
Aluminum All Mettler Toledo Moisture Analyzers Disposable Pans - Pk. of 80
Case of 80 Each for $360.44
Edge
Description
Description
Choice of washable, reusable stainless chrome-nickel steel pans for halogen moisture analyzers, or aluminum or stainless-steel pans for all Mettler Toledo moisture analyzers.
  • 3.75 in. (96mm) in diameter
Specifications
Specifications
Round
Moisture Pan
All Mettler Toledo Moisture Analyzers
88.9 mm
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'