Mettler Toledo™ Bluetooth RS232 Serial Adapters for P50 Line Thermal Printers
For use with P50 Line Thermal Printers.
Supplier: Mettler Toledo™ 30086494
Description
Bluetooth RS232 Serial Adapter
- Accessory for P-56RUE and P-58RUE Thermal printers
- For wireless connection between printer and balance or between balance and PC
- Single or paired
Specifications
P-56RUE and P-58RUE Thermal Printers | |
Single | |
Bluetooth RS232 Serial Adapter |