Mettler Toledo™ perfectION™ Combination Electrodes
Enables optimum contact between the electrolyte and measurement solution
Supplier: Mettler Toledo™ 51344851
Description
- Electrolyte solution flows easily through junction by pressing electrode head downwards
- Ensures that junction is optimally cleaned between measurements
- Electrolyte can be simply emptied and refilled if necessary
- Sleeve junction that enables optimum contact between the electrolyte and measurement solution, even in difficult samples
- Consist of a measuring electrode complete with an integrated reference
- Sensor with attached cable and BNC connector
- Reference electrolyte (60 mL)
- Operating instructions
Specifications
Can flow easily through the junction and junction is optimally cleaned between the measurements and the electrolyte can be simply emptied and refilled | |
ISA | |
perfectION membrane module K |
perfectION membrane module K | |
32° to 104°F | |
0°C to 40°C |