Mettler Toledo™ Replacement Parts for V20 Karl Fischer™ Autotitrator
For use with V20 Karl Fischer Autotitrators.
Supplier:  Mettler Toledo™ 23961
 View more versions of this product
Catalog No. 36-101-3094
Explore available promotions
Description
Description
  • Accessory for DV1000 titration burettes
  • For titrant protection from moisture or CO2
  • With cover
Specifications
Specifications
Drying Cup
Water content determination
453.5 g