Mettler Toledo™ Expansion Modules for SevenExcellence™ Meters
The modular design of SevenExcellence allows you to expand your instrument with additional measurement parameters for future needs at any time.
Supplier:  Mettler Toledo™ 30034475
 View more versions of this product
Catalog No. 01-911-603
Explore available promotions
Description
Description
Just add the new expansion unit, switch on the instrument and the new expansion unit is automatically recognized and ready to use! SevenExcellence fits in total three expansion units in any composition.
Specifications
Specifications
Blank Module