Mettler Toledo™ Expansion Modules for SevenExcellence™ Meters
The modular design of SevenExcellence allows you to expand your instrument with additional measurement parameters for future needs at any time.
Supplier: Mettler Toledo™ 30034473
Description
Just add the new expansion unit, switch on the instrument and the new expansion unit is automatically recognized and ready to use! SevenExcellence fits in total three expansion units in any composition.
Specifications
±0.1°C | |
0.001μS/cm to 2000mS/cm | |
0.001mg/L to 1000g/L | |
0.1°C | |
SevenExcellence Benchtop Meters |
Conductivity Module | |
0 to 80psu | |
±0.5% | |
-30°C to +130°C |