METTLER TOLEDO™ SevenExcellence™ S470 pH/Conductivity Benchtop Meter
The S470 meter measures pH, conductivity, resistivity, redox/ORP, TDS, salinity, and ISFET. It can be expanded with additional modules to measure dissolved oxygen, BOD, and ISE. It has an intuitive touchscreen, measurement shortcuts, and can export data to a printer, Excel file, or LIMS system.
Supplier:  METTLER TOLEDO™ 30046254
 View more versions of this product
Catalog No. 01-910-913
Explore available promotions
Description
Description
  • pH, conductivity, resistivity, redox/ORP, TDS, salinity, and ISFET meter than can be expanded with additional measurement modules to measure dissolved oxygen, ISE, and BOD
  • Easily create measurement and calibration shortcuts
  • 7 inch color touch screen with intuitive menu navigation
  • Compatible with a wide range of electrodes for any application
  • Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
  • Meter connects to electrodes via BNC; S7 to BNC cables available
  • Export data to a printer, Excel file, or LIMS system
  • Automation available for measurements and cleaning in high-throughput applications
  • Manage user permissions and access to ensure data integrity
  • Compatible with LabX software for advanced data management and 21CFR part 11 regulatory compliance
  • Optional comprehensive qualification package available including IQ/OQ/PQ
Specifications
Specifications
RS232, USB-A, USB-B, LAN, Mini-LTW, Mini-DIN, BNC, Cinch/RCA
±0.1mV
13 Pre defined and 20 user defined standard
Max. 5 points, 8 pre defined and 20 user defined buffer group
TFT
-2000 to 2000 mV
0.01 to 100 Mohm/cm
0.001 mg/L to 1000 g/L
0.001/0.01/0.1
S470 pH/conductivity meter, InLab Pure Pro-ISM pH electrode, InLab 741-ISM conductivity electrode(1.2 m cable), electrode holder, 2 buffer sachets each for pH 4.01, 7, 9.21, 10.01, EasyDirect pH activation key, test certificate, pH measurement guide, conductivity/DO measurement guide, multipin-BNC/RCA electrode cable 1.2 m
0 to 999999mg/L
0.1/1mV
0.001 to 1
±0.1°C
±0.5%
±0.002
Max. 5 points, 8 pre defined and 20 user defined buffer group
20,000
0.001 μS/cm to 2000 mS/cm
-2 to 20
0 to 80 ppt
0.001 to 1
-30°C to +130°C
S470 Kit - USP/EP Compliant
0.001μS/cm to 2000mS/cm
0.001 to 1
0.001 to 1
0.1°C