METTLER TOLEDO™ Portable pH/Conductivity/Dissolved Oxygen/Ion Meters
Water and dust proof, IP67 rated meter for portable or field measurements. Portable meters compatible with pH, conductivity, dissolved oxygen, and ion selective electrodes.
Supplier:  METTLER TOLEDO™ 51302600
 View more versions of this product
Catalog No. 01-917-130
Explore available promotions
Description
Description
  • Dust proof
  • Water proof
  • IP67 Rated
  • pH, Redox/ORP, conductivity, dissolved oxygen and ISE measuring capability (model dependent)
  • Compatible with a wide range of IP67-rated electrodes for any application
  • Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
  • Data storage available; capacity dependent on model
  • Export data to a printer or Excel file for Seven2Go Pro only
  • EasyDirect software available for data transfer (model dependent) for Seven2Go Pro only
  • Rubber holster available for single-handed measurements
  • uGo carrying and protecting case included in selective kits (see below for complete list); available for all portable meters
Specifications
Specifications
±0.5%
±0.01
4 x AA
ISO 9001, ISO 14001
LCD
0.01 μS/cm to 500 mS/cm
0 to 14
0 to 80 ppt
0.01
Meter Only
Dual Channel; pH/ORP, conductivity
Meter only (SG23)
1mV
±0.5°C
0°C to 40°C
±1mV
±0.5%
3 Points, 4 pre defined buffer group
99 GLP data records
IP 67
-1999 to 1999 mV
0 to 100 Mohm/cm
0.01 mg/L to 300 g/L
Determination of pH, mV, conductivity, TDS (total dissolved solids), salinity and specific resistance
pH: BNC/RCA, conductor: LTW 7-pin
NTC
0.01/1
0.01/1
30°F to 104°F
0.1°C