missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
METTLERÂ TOLEDO™ Portable pH/Conductivity/Dissolved Oxygen/Ion Meters
Water and dust proof, IP67 rated meter for portable or field measurements. Portable meters compatible with pH, conductivity, dissolved oxygen, and ion selective electrodes.
Supplier: METTLERÂ TOLEDO™ 51302620
Description
- Dust proof
- Water proof
- IP67 Rated
- pH, Redox/ORP, conductivity, dissolved oxygen and ISE measuring capability (model dependent)
- Compatible with a wide range of IP67-rated electrodes for any application
- Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
- Data storage available; capacity dependent on model
- Export data to a printer or Excel file for Seven2Go Pro only
- EasyDirect software available for data transfer (model dependent) for Seven2Go Pro only
- Rubber holster available for single-handed measurements
- uGo carrying and protecting case included in selective kits (see below for complete list); available for all portable meters
Specifications
IR to printer or PC via RS232 or USB | |
±0.1mV | |
±0.5% | |
5 Points, 7 pre-defined buffer group | |
LCD Graphic, Backlit | |
0.01 μS/cm to 1000 mS/cm | |
-2 to 19.999 | |
0 to 80 ppt | |
0.1/0.01/0.001 | |
Meter Only | |
Dual Channel;Â pH/ORP/ion, conductivity | |
Portable Meter | |
0.1mV | |
±0.2°C |
±0.5% | |
±0.002 | |
4 x AA | |
ISO 9001, ISO 14001 | |
IP 67 | |
-1999 to 1999 mV | |
0 to 100 Mohm/cm | |
0.01 mg/L to 300 g/L | |
Determination of pH, mV, rel. mV or ion concentrations and conductivity, TDS (total dissolved solids), salinity or specific resistance | |
pH: BNC/RCA, conductor: LTW 7-pin | |
NTC | |
0.01/1 | |
0.01/1 | |
0.1°C |