Mettler Toledo™ Tweezers for Analytical and Calibration Weights
Accessories for Weights. Ergonomic Weight Handling - Ergonomic tweezers and weight forks are made of clean room suitable materials and allow professional weight handling and balance testing.
Supplier:  Mettler Toledo™ 15901
 View more versions of this product
Catalog No. 01-915-379
Explore available promotions
Description
Description
Constructed of metal with carbon reinforced nylon.
Specifications
Specifications
Tweezers
Metal with Carbon Reinforced Nylon
1g-1kg Weights
Each