missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MFG Tray Chemical-Resistant Multipurpose Trays and Pans

Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills

Supplier:  MFG Tray 3034031537

 View more versions of this product

Catalog No. 15-239-4


Explore available promotions

Description

Description

Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills

  • Reinforced white thermoset polyester composite construction Resistant to most detergents, mild acid and alkaline solutions, corrosive salts and many reagents
  • Withstand temperatures from -51° to +121°C (-60° to +250°F) continuous exposure, or 149°C (300°F) intermittent
  • High load-bearing capacity
  • Excellent dimensional stability Easily steam cleaned

Order Info

Available in 1 in. deep (2.5cm) trays, shallow pans from 1 1/4 to 2 3/4 in. deep (3 to 7cm), and ventilated stacking pans from 2 3/4 to 4 1/8 in. deep (7 to 10.5cm).

Specifications

Specifications

13.25 x 10.62 x 1 in. (34 x 27 x 2.5 cm)
2.5 cm
Fiberglass
27 cm
13.25 in.
Rectangular
-51°C to +121°C
White
34 cm
Multi-Purpose Tray
Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills
1 in.
-60°F to +250°F
10.62 in.
Videos
SDS
Documents

Documents

Promotions

Promotions