missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MFG Tray Chemical-Resistant Multipurpose Trays and Pans
Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills
Supplier: MFG Tray 3044031537
Description
Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills
- Reinforced white thermoset polyester composite construction Resistant to most detergents, mild acid and alkaline solutions, corrosive salts and many reagents
- Withstand temperatures from -51° to +121°C (-60° to +250°F) continuous exposure, or 149°C (300°F) intermittent
- High load-bearing capacity
- Excellent dimensional stability Easily steam cleaned
Order Info
Available in 1 in. deep (2.5cm) trays, shallow pans from 1 1/4 to 2 3/4 in. deep (3 to 7cm), and ventilated stacking pans from 2 3/4 to 4 1/8 in. deep (7 to 10.5cm).
Specifications
20.38 x 15.12 x 1 in. (52 x 38.5 x 2.5 cm) | |
2.5 cm | |
Fiberglass | |
38.5 cm | |
20.38 in. | |
Rectangular | |
-51°C to +121°C |
White | |
52 cm | |
Multi-Purpose Tray | |
Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills | |
1 in. | |
-60°F to +250°F | |
15.12 in. |