Intertek™ MIC Test Kit, Petroleum Microbiology Media: Acid Producing Bacteria Media DFS Item
Microbiology growth media products
Supplier:  Intertek™ MK106C
 View more versions of this product
Catalog No. 08629021C
Explore available promotions
Description
Description
A range of bacterial growth media and accessories for use in detecting and quantifying microbiologically influenced corrosion (MIC). Media products are available as boxes of 12 vials, cases of 288 vials, and in the single tube quick-test format.
Acid Producing Bacteria (APB) contribute to MIC by decreasing the pH of the environment which promotes the dissolution of metal.
  • APB: for quantification of acid producing bacteria and general heterotrophic anaerobic bacteria
  • The change in pH (production of acid) in APB-Q media is indicated by a change in color from red to yellow
Compliance
Manufactured in accordance with NACE Standard TM0194-94, Field Monitoring of Bacterial Growth in Oilfield Systems.
Specifications
Specifications
NACE, TM0194-94
United States
Cultivation of Acid producing bacteria, MIC testing
12 Vials
Red
MIC Test Kit
Vial