Intertek™ MIC Test Kit, Petroleum Microbiology Media: Iron Reducing Bacteria Media DFS Item
Microbiology growth media products
Supplier:  Intertek™ MK116B
 View more versions of this product
Catalog No. 08629011B
Explore available promotions
Description
Description
A range of bacterial growth media and accessories for use in detecting and quantifying microbiologically influenced corrosion (MIC). Media products are available as boxes of 12 vials, cases of 288 vials, and in the single tube quick-test format.
Iron Reducing Bacteria (IRB) transform ferric iron to ferrous iron and are involved with slime formation, odor and water discoloration to brown or black.
  • IRB: for quantification of iron reducing bacteria
  • IRB: Q quick format single tube test for IRB
  • The IRB-Q media provides a convenient way to estimate the concentration of IRB in water samples by changing the color of the media from light golden to brown or black, and the clarity of the media changes from clear to turbid/cloudy
Compliance
Manufactured in accordance with NACE Standard TM0194-94, Field Monitoring of Bacterial Growth in Oilfield Systems.
Specifications
Specifications
NACE, TM0194-94
MIC Test Kit
Vial
United States
Cultivation of Iron reducing bacteria, MIC testing
288 Vials