missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Michler's Ketone, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1188936

 View more versions of this product

Catalog No. AAA1188936


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Michler's Ketone
174°C to 176°C
220°C (428°F)
MFCD00008312
790733
michler's ketone, 4,4'-bis dimethylamino benzophenone, michler ketone, bis 4-dimethylamino phenyl methanone, p,p'-michler's ketone, michlers ketone, methanone, bis 4-dimethylamino phenyl, tetramethyldiaminobenzophenone, p,p'-bis dimethylamino benzophenone
CN(C)C1=CC=C(C=C1)C(=O)C1=CC=C(C=C1)N(C)C
268.36
CHEBI:82347
98%
90-94-8
>360°C
C17H20N2O
500 g
14,6177
VVBLNCFGVYUYGU-UHFFFAOYSA-N
bis[4-(dimethylamino)phenyl]methanone
7031
268.36

Chemical Identifiers

90-94-8
268.36
VVBLNCFGVYUYGU-UHFFFAOYSA-N
7031
bis[4-(dimethylamino)phenyl]methanone
C17H20N2O
MFCD00008312
michler's ketone, 4,4'-bis dimethylamino benzophenone, michler ketone, bis 4-dimethylamino phenyl methanone, p,p'-michler's ketone, michlers ketone, methanone, bis 4-dimethylamino phenyl, tetramethyldiaminobenzophenone, p,p'-bis dimethylamino benzophenone
CHEBI:82347
CN(C)C1=CC=C(C=C1)C(=O)C1=CC=C(C=C1)N(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H350-H341-H318
May cause cancer.
Suspected of causing genetic defects.
Causes serious eye damage.

P201-P202-P281-P305+P351+P338-P308+P313-P310-P501c

H318-H341-H350

EINECSNumber : 202-027-5

RTECSNumber : DJ0250000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only