Micro Essential Lab Hydrion™ Spectral Plastic pH Strips
Single color match — just dip and read
Supplier:  Micro Essential Lab 9600
 View more versions of this product
Catalog No. 14-850-112
Explore available promotions
Description
Description
  • Economical — low cost per test
  • Accurate — same high-quality pH test in plastic strip format
  • Convenient — packed in handy flip-top vials
  • Made in the USA
Specifications
Specifications
6.5 to 13
Colorimetric chart
pH Test Strip
0.5
Each
Product Suggestions
Product Suggestions